Skip to Content
Merck
All Photos(2)

Key Documents

MABN369

Sigma-Aldrich

Anti-Neuroglobin Antibody, clone 6G1.1

clone 6G1.1, from mouse

Synonym(s):

Neuroglobin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€432.00

€432.00


Estimated to ship onApril 10, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€432.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€432.00


Estimated to ship onApril 10, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G1.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NGB(58157)

General description

Neuroglobin protein encoded by the gene NGB, is involved in oxygen transport in the brain. Neuroglobin is also important for neuroprotection during oxidative stress. Additionally, Neuroglobin has anti-apoptotic activity by reseting the mitochondrial membrane tigger level for cytochrome C release and the commitment to apoptosis. Neuroglobin is localized to the perikaryon, cytoplasm, and mitochondrion. Neuroglobin is predominantly expressed in the brain with strongest expression in the frontal lobe and the subthalamic nucleus and thalamus.

Immunogen

GST-tagged recombinant protein corresponding to the N-terminus of human Neuroglobin.

Application

Detect Neuroglobin using this mouse monoclonal antibody, Anti-Neuroglobin Antibody, clone 6G1.1 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Neuroglobin in human thalamus and human frontal lobe tissue.

Quality

Evaluated by Western Blotting in human cortex tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Neuroglobin in 10 µg of human cortex tissue lysate.

Target description

~17 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrizio Cracco et al.
International journal of molecular sciences, 24(6) (2023-03-30)
A promising therapeutic strategy to delay and/or prevent the onset of neurodegenerative diseases (NDs) could be to restore neuroprotective pathways physiologically triggered by neurons against stress injury. Recently, we identified the accumulation of neuroglobin (NGB) in neuronal cells, induced by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service