Skip to Content
Merck
All Photos(3)

Key Documents

MABN361

Sigma-Aldrich

Anti-VAPA Antibody, clone 7E10.1

clone 7E10.1, from mouse

Synonym(s):

Vesicle-associated membrane protein-associated protein A, VAMP-A, VAMP-associated protein A, VAP-A, 33 kDa VAMP-associated protein, VAP-33, VAPA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€395.00

€395.00


Estimated to ship onApril 23, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€395.00


Estimated to ship onApril 23, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7E10.1, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VAPA(9218)

General description

Vesicle-associated membrane protein-associated protein A (UniProt Q9P0L0; also known as 33 kDa VAMP-associated protein, hVAP-33, VAMP-A, VAMP-associated protein A, VAP-33, VAP-A) is encoded by the VAPA (also known as VAP33) gene (Gene ID 9218) in human. VAMP-associated proteins (VAPs) are type IV membrane proteins that are well conserved among species. There exist three human VAPs encoded by two genes, with VAPA encoding VAP-A and VAPB encoding VAP-B and VAP-C. VAPs generally localize at the endoplasmic reticulum (ER), although they are also reported to localize at other subcellular organelles in some species and cell types. VAP-A contains a major sperm protein (MSP) domain at the N-terminal, followed by a coiled-coil domain, and a transmembrane (TM) domain. VAPs were shown to have important roles in non-vesicular lipid transport, lipid metabolism, the regulation of ER structure, and the unfolded protein response through MSP domain-mediated interaction with FFAT motifs. Oxysterol-binding protein (OSBP) is a cytosolic receptor of cholesterol and oxysterols. OSBP is recruited to the ER by binding to the MSP domain of VAP-A, a process essential for the stimulation of sphingomyelin synthesis by 25-hydroxycholesterol.

Specificity

Cross-reactivity to human VAPA spliced isoform 2 is highly expected, but not confirmed.

Immunogen

Epitope: C-terminal half
GST-tagged recombinant protein corresponding to the C-terminal half of human VAPA.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-VAPA Antibody, clone 7E10.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of VAPA.
Western Blotting Analysis: 0.2 µg/mL from a representative lot detected VAPA in 10 µg of mouse brain tissue lysate.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected VAPA in human cerebellum and human testis tissue.

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected VAPA in 10 µg of SH-SY5Y cell lysate.

Target description

~30 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service