MABN2276
Anti-Dab1 Antibody, clone 4E12
clone 4E12, from rat
Synonym(s):
Disabled homolog 1
Select a Size
Select a Size
About This Item
biological source
rat
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
4E12, monoclonal
species reactivity
mouse
packaging
antibody small pack of 25 μL
technique(s)
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
mouse ... Dab1(13131)
General description
Specificity
Immunogen
Application
Neuroscience
Immunoprecipitation Analysis: A 1:500 dilution from a representative lot immunoprecipitated Dab1 in cerebral cortex tissue lysate from P20 mouse (Courtesy of Prof. Mitsuharu Hattori, Nagoya City University).
Immunohistochemistry Analysis: A representative lot detected Dab1 in Immunohistochemistry applications (Onoune, A., et. al. (2014). Neurosci Res. 88:23-27).
Quality
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Dab1 in lysates from HEK293T cells expressing murine Dab 1.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service