Skip to Content
Merck
All Photos(2)

Key Documents

MABN1583

Sigma-Aldrich

Anti-CALML5 Antibody, clone 3F6.1

clone 3F6.1, from mouse

Synonym(s):

Calmodulin-like protein 5, Calmodulin-like skin protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€276.00

€276.00


Estimated to ship onMarch 26, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€276.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€276.00


Estimated to ship onMarch 26, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F6.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CALML5(51806)

General description

Calmodulin-like protein 5 (UniProt Q9NZT1; also known as Calmodulin-like skin protein) is encoded by the CALML5 (also known as CLSP) gene (Gene ID 51806) in human. Epidermal differentiation and barrier function depend on many calcium-regulated proteins and enzymes. CALML5/CLSP is expressed in the stratum granulosum and lower layers of the stratum corneum of the skin, where it modulates the function of calcium-dependent proteins, such as transglutaminase 3. CALML5/CLSP level is upregulated in granular and spinal keratinocytes during the later stages of wound healing in human epidermis and eventually returns to basal level once the wound is closed and barrier function has been restored. In the hyperproliferative skin disorders psoriasis and atopic dermatitis (AD), the permeability barrier is disturbed, causing an increased CLSP built-up as a result of decresed CLSP degradation due to an altered calcium gradient.

Specificity

Clone 3F6.1 recognizes an epitope outside the first EF-hand motif.

Immunogen

Epitope: Outside the first EF-hand motif.
GST-tagged recombinant human CALML5 lacking the first EF-hand motif.

Application

Anti-CALML5 Antibody, clone 3F6.1 is an antibody against CALM5 for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected CALML5 in human tonsil and skin tissue sections.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting of human CALML5 GST fusion protein.

Western Blotting Analysis: 0.05 µg/mL of this antibody detected 0.05 µg of human CALML5 GST fusion protein.

Target description

15.89 kDa calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service