Skip to Content
Merck
All Photos(2)

Documents

MABF3175

Sigma-Aldrich

Anti-IL-5 Antibody, clone 4A1.6.15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4A1.6.15, monoclonal

mol wt

calculated mol wt 15.24 kDa
observed mol wt ~226 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... IL5(3567)

Related Categories

General description

Interleukin-5 (UniProt: P05113; also known as IL-5, B-cell differentiation factor I, Eosinophil differentiation factor, T-cell replacing factor, TRF) is encoded by the IL5 gene (Gene ID: 3567) in human. IL-5 is a disulfide-linked homodimeric secreted protein that is synthesized and secreted mainly by the activated CD4+ helper T cells of the Th2 subset, mast cells, and eosinophils. It is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to produce the mature protein (aa 20-134). It serves as a powerful proinflammatory cytokine that is responsible for maturation, proliferation, activation, and migration of eosinophils. It is also a key factor in the upregulation of myeloid progenitors in the bone marrow after airway allergen challenge, and for trafficking from the marrow to the airways in upper or lower airways inflammation. Its biological effects are mediated via selective interaction with IL-5 receptor (IL-5R), which is composed of an a subunit (IL-5Ra) and a non-specific βc heterodimer. As a homodimer it binds to IL-5Ra expressed on eosinophil surface and recruits the βc dimer and induces the assembly of the IL-5/IL-5Ra/βc ternary complex. In the absence of IL-5, IL-5Ra is complexed with the intracellular JAK2, whereas the βc subunit is associated with JAK1. (Ref.: Pelaia, C., et al. (2019). Front. Physiol. 10; 1514).

Specificity

Clone 4A1.6.15 is a mouse monoclonal antibody that detects human Interleukin-5 (IL-5).

Immunogen

Full-length recombinant human Interleukin-5 (without the signal peptide).

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant human IL-5.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant human IL-5.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected IL-5 in human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-IL-5, clone 4A1.6.15, Cat. No. MABF3175, is a mouse monoclonal antibody that detects human Interleukin-5 and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service