MABF2760
Anti-CD200/OX2 Antibody, clone OX-90

Synonym(s):
MRC OX-2 antigen
About This Item
biological source
rat
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
OX-90, monoclonal
mol wt
calculated mol wt 31.26 kDa
purified by
using protein G
species reactivity
mouse
packaging
antibody small pack of 100
technique(s)
ELISA: suitable
blocking: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
isotype
IgG2aκ
epitope sequence
Extracellular domain
UniProt accession no.
storage temp.
-10 to -25°C
Gene Information
mouse ... Cd200(17470)
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in BALB/c mouse splenocytes.
Flow Cytometry Analysis: 1 µg from a representative lot detected CD200/OX2 in one million BALB/c mouse splenocytes.
Tested Applications
Flow Cytometry Analysis: A representative lot detected CD200/OX2 in flow cytometry application (Hoek, R.M., et al. (2000). Science. 290(5497):1768-71; Broderick, C., et al. (2002). Am J Pathol. 161(5):1669-77; Akkaya, M., et al. (2013). PLoS One. 8(5):e63325).
Enzyme Immunoassay (ELISA): A representative lot detected CD200/OX2 in ELISA application (Hoek, R.M., et al. (2000). Science. 290(5497):1768-71).
Immunofluorescence Analysis: A representative lot detected CD200/OX2 in immunofluorescence application (Broderick, C., et al. (2002). Am J Pathol. 161(5):1669-77; Snelgrove, R.J., et al. (2008). Nat Immunol. 9(9):1074-83).
Blocking of Interfering Antibodies: A representative lot effectively blocked endogenous binding of CD200/OX2 to CD200R (Snelgrove, R.J., et al. (2008). Nat Immunol. 9(9):1074-83; Akkaya, M., et al. (2013). PLoS One. 8(5):e63325).
Immunohistochemistry Analysis: A representative lot detected CD200/OX2 in immunohistochemistry application (Hoek, R.M., et al. (2000). Science. 290(5497):1768-71; Broderick, C., et al. (2002). Am J Pathol. 161(5):1669-77; Snelgrove, R.J., et al. (2008). Nat Immunol. 9(9):1074-83).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service