MABE1153
Anti-MRE11 Antibody, clone 15B8.1E7.6
clone 15B8.1E7.6, from hamster(Armenian)
Synonym(s):
Double-strand break repair protein MRE11A, MmMRE11A, Meiotic recombination 11 homolog 1, MRE11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog A
Select a Size
About This Item
Recommended Products
biological source
hamster (Armenian)
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
15B8.1E7.6, monoclonal
species reactivity
mouse, human
packaging
antibody small pack of 25 μg
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... MRE11(4361)
General description
Specificity
Immunogen
Application
Epigenetics & Nuclear Function
Quality
Western Blotting Analysis: 4 µg/mL of this antibody detected MRE11 in 10 µg of Ramo cell lysate.
Target description
Linkage
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service