Skip to Content
Merck
All Photos(2)

Key Documents

MAB1501-AF647

Sigma-Aldrich

Anti-Actin Antibody, clone C4, Alexa Fluor 647 conjugate

clone C4, from mouse, ALEXA FLUOR 647

Synonym(s):

Actin, alpha skeletal muscle, Alpha-actin-1, Actin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€485.00

€485.00


Estimated to ship onApril 18, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€485.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€485.00


Estimated to ship onApril 18, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified antibody

antibody product type

primary antibodies

clone

C4, monoclonal

species reactivity

human

species reactivity (predicted by homology)

chicken (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)

General description

Actins are ubiquitous eukaryotic proteins that serve as a multi-functional, basic building blocks of cytoskeletal microfilaments. They play critical roles in a wide range of cellular processes, including cell division, cell migration, chromatin remodeling, trascriptional regulation and vesicle trafficking. These funstions are attributed to their ability to form filaments, which can quickly assemble and disassemble depending upon the needs of the cell. At least six different actin types have been reported in mammals. Although actins show about 90% overall sequence homology, isoforms do not show spatial, temporal and tissue-specific expression patterns and only 50-60% homology is found in their 18 N-terminal residues. Beta and gamma-actins, also known as cytoplasmic actins, are highly conserved in higher animals and are predominantly expressed in non-muscle cells where they control cell structure. Exocytosis, and motility. They are nearly identical proteins and differ only in four amino acids at the N-terminal region. The other four actin isoforms are typically found in specific adult muscle tissue types. Alpha-cardiac and alpha-skeletal actins are expressed in striated cardiac and skeletal muscles, respectively. Alpha and gamma actins are primarily found in vascular smooth muscle and enteric smooth muscles, respectively. It has been shown that under calcium-bound conditions, beta-actin exhibits more dynamic behavior than gamma-actin with higher rates of polymerization and depolymerization. Also, beta- and gamma-actins can readily copolymerize, and the resulting filaments exhibit polymerization and depolymerization rates that vary depending on the ration of beta- to gamma-actin (Lessard, JL.,et al.(1988). Cell Motility Cytoskeleton 10(3); 349-362.

Specificity

To date, all animal species and cell types with an actin form react by indirect immunofluorescence or immunoblot, including plant actin.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Actin in NIH/3T3 cells.
The unconjugated version of this antibody (Cat. No. MAB1501) has been demonstrated to perform in WB, ELISA, ICC, IF, IHC, IH(P).
This Anti-Actin Antibody, clone C4, Alexa Fluor 647 conjugate is validated for use in Immunocytochemistry for the detection of Actin.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Actin in HeLa cells.

Target description

40 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service