Skip to Content
Merck
All Photos(2)

Key Documents

HTS170M

Sigma-Aldrich

ChemiSCREEN Membrane Preparation Recombinant Human sst1 Somatostatin Receptor

Human sst1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
€697.00

€697.00


Estimated to ship onApril 28, 2025Details



Select a Size

Change View
200 UNITS
€697.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

€697.00


Estimated to ship onApril 28, 2025Details


biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... SSTR1(6751)

General description

Human sst1
Somatostatin, a 14 or 28 amino acid peptide, inhibits hormone secretion from exocrine glands, and displays an antiproliferative activity on a number of cell types. A family of 5 Gi-coupled receptors mediates the effects of somatostatin. The classical activity of somatostatin of inhibiting growth hormone release from the pituitary appears to be mediated by sst1. In addition, sst1 functions as an inhibitory autoreceptor on somatostatin neurons in the hippocampus, hypothalamus, basal ganglia and retina (Thermos et al., 2006). The somatostatin receptors are targets for anticancer drugs and for treatment of hormonal dysfunction. Millipore′s sst1 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of agonists and antagonists of sst1. The membrane preparations exhibit a Kd of 1.4 nM for [125I]- somatostatin-14. With 0.5 nM [125I]- somatostatin-14, 5 μg/well sst1 Membrane Prep yields greater than 12 fold signal-to-background ratio.

Application

Human sst1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Radioligand binding assay and GTPγS binding

Biochem/physiol Actions

GPCR Class: A
Protein Target: sst1
Target Sub-Family: Somatostatin

Quality

Table 1. Signal:background and specific binding values obtained in a competition binding assay with varying amounts of sst1 membrane prep.
5 µg/well
Signal:Background 20.4
Specific Binding (cpm) 14,647

SPECIFICATIONS: 1 unit = 5 μg membrane preparation
Bmax: 8.4 pmol/mg
Kd: 1.4 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM HEPES, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I]- somatostatin-14 (PerkinElmer NEX389)
Wash Buffer: 50 mM HEPES, pH 7.4, 500 mM NaCl, 0.1% BSA, filtered and stored at 4°C.

Physical form

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 12-fold signal:background with [125I]- somatostatin-14 at 0.5 nM.

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membrane protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Store at –70°C. Product is stable for at least 6 months from the date of receipt when stored as directed. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service