Skip to Content
Merck
All Photos(1)

Key Documents

GF319

Sigma-Aldrich

BMP-2, Human Recombinant
Animal Free

Synonym(s):

BMP2, BMP 2A, BMP 2, BMP2A, bone morphogenetic protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€392.00

€392.00


Estimated to ship onApril 18, 2025Details



Select a Size

Change View
10 μG
€392.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

€392.00


Estimated to ship onApril 18, 2025Details


impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

dry ice

General description

BMP-2 is a potent osteoinductive cytokine, capable of inducing bone and cartilage formation in association with an osteoconductive carrier such as collagen and synthetic hydroxyapatite. In addition to its osteogenic activity, BMP-2 appears to play an important role in cardiac morphogenesis, and is expressed in a variety of other tissues including lung, liver, spleen, prostate, ovary, and small intestine. Recombinant human BMP-2 is a 26.0 kDa homodimeric disulfide-linked protein consisting of two identical 115 amino acid chains.

Quality

Bioactivity assay: Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 0.5-1.0 μg/ml.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service