Skip to Content
Merck
All Photos(1)

Documents

GF315

Sigma-Aldrich

VEGF165 Protein, Human Recombinant Animal Free

The Vascular endothelial growth factor (VEGF or VEGFA) is a potent mediator of both angiogenesis & vasculogenesis in the fetus & adult. Manufactured using all non-animal reagents.

Synonym(s):

Vascular Endothelial Growth Factor 165, VEGF-165, VEGF, Vascular Endothelial Growth Factor, VEGF-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

Quality Level

Assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

UniProt accession no.

shipped in

dry ice

General description

Product Source: E. coli
Vascular endothelial growth factor (VEGF or VEGFA) is a potent mediator of both angiogenesis and vasculogenesis in the fetus and adult. Humans express alternately spliced isoforms of 121, 145, 165, 183, 189, and 206 amino acids (aa) in length. VEGF165 appears to be the most abundant and potent isoform, followed by VEGF121 and VEGF189. VEGF is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. Substantial evidence implicates VEGF in the induction of tumor metastasis and intra-ocular neovascular syndromes. VEGF signals through the three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant human VEGF is a 38.2 kDa disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains.

Specificity

Cross Reactivty
Mouse, Rat, Pig, Hamster, Trout, Green Algae, Sheep, Cow, Rabbit, Guinea Pig

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

Bioactivity assay: Determined by the dose-dependent stimulation of the proliferation of human umbilical vein endothelial cells (HUVEC) using a concentration range of 1.0-8.0 ng/ml.

Physical form

Product is filtered through a 0.2 micron filter before lyophilization.
Recombinant animal free human VEGF165 is manufactured using all non-animal reagents

Storage and Stability

Store at -20°C for up to 6 months from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John P Morgan et al.
Nature protocols, 8(9), 1820-1836 (2013-08-31)
This protocol describes how to form a 3D cell culture with explicit, endothelialized microvessels. The approach leads to fully enclosed, perfusable vessels in a bioremodelable hydrogel (type I collagen). The protocol uses microfabrication to enable user-defined geometries of the vascular
Wutigri Nimlamool et al.
Molecular biology of the cell, 26(15), 2755-2768 (2015-06-13)
Gap junctions (GJs) exhibit a complex modus of assembly and degradation to maintain balanced intercellular communication (GJIC). Several growth factors, including vascular endothelial growth factor (VEGF), have been reported to disrupt cell-cell junctions and abolish GJIC. VEGF directly stimulates VEGF-receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service