Skip to Content
Merck
All Photos(1)

Key Documents

AB5604

Sigma-Aldrich

Anti-Calcium Channel Antibody, Voltage Gated α 2/δ-1, L-type

Chemicon®, from rabbit

Synonym(s):

Cacna2d1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CACNA2D1(781)

Specificity

Recognizes alpha2/delta-1 subunit, N-terminal. The epitope does not share homology with any other known proteins. Reactivity has been verified only in rat membranes.

Immunogen

Epitope: Voltage Gated alpha 2/delta-1, L-type
Peptide corresponding to 15 amino acids from the rabbit alpha2 subunit (residues 27-41 of rabbit alpha2/delta-1 precursor) P13806. By sequence comparsions, the immunogen sequence used is identical in human, porcine, with mouse and rat missing only a single amino acid.

Application

Detect Calcium Channel using this Anti-Calcium Channel Antibody, Voltage Gated α 2/δ-1, L-type validated for use in WB.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 20 μg of control antigen. The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 0.75 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service