Skip to Content
Merck
All Photos(1)

Documents

AB1094

Sigma-Aldrich

Anti-Myosin X Antibody

serum, from rabbit

Synonym(s):

Unconventionnal myosin-X, Unconventional myosin-10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

GenBank accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYO10(4651)

General description

Myosins are a superfamily of molecular motor proteins found in eukaryotic cells. Motor proteins are involved in muscular contraction, cytokinesis, short-range membrane/vesicle transport and a host of other cellular processes. The myosins are responsible for actin-based motility by using the energy of ATP hydrolysis to generate force and movement along actin filaments. There are many isoforms of myosin of which some have specialized functions in certain cell types (such as muscle), while other isoforms are ubiquitous. Myosin X belongs to a group of myosin isoforms known as unconventional myosins that participate in cellular processes ranging from membrane trafficking to signaling and cell motility. Myosin X is concentrated at the tips of filopodia and Myosin X rich regions travel forward and backward along filopods, (Berg, 2002). It also appears that Myosin X functions in actual filopodia formation, (Bohil, 2006).

Specificity

Cat. No. AB1094 recognizes human Myosin X.
Predicted to cross react with mouse and rat based on 100% sequence homology. Reactivity with other species has not been tested.

Immunogen

Epitope: a.a. 936-953
KLH-conjugated synthetic peptide to amino acids 936-953 of human Myosin X.

Application

Detect Myosin X using this Anti-Myosin X Antibody validated for use in WB.

Quality

Evaluated on a representative lot by Western blot on human head/neck lysate using Anti-Myosin X.

Target description

~ 108 kDa

Storage and Stability

Maintain at -20°C for up to 1 year from date of receipt.
Upon arrival, aliquot into usable quantities to avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Human head/neck lysate.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service