Skip to Content
Merck
All Photos(1)

Key Documents

964204

Supelco

Display and alarm device for bottle level sensor

Synonym(s):

Display level sensor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

shipped in

ambient

General description

The display is the completion for our bottle level sensors 9.67100.2001 and 9.67100.2002. It is equipped with 4 diodes for 4 alarm points, e.g. 4 different bottle sensors. If the level limit is reached, the diodes start to flash and a acoustical signal can be heard clearly. By pushing the reset button the signals can be confirmed. We recommend an easily visible position at the workplace for mounting.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service