Skip to Content
Merck
All Photos(1)

Documents

8.14179

Sigma-Aldrich

Disulfur dichloride

for synthesis

Synonym(s):

Disulfur dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2S2
CAS Number:
Molecular Weight:
135.04
MDL number:
UNSPSC Code:
12352307
EC Index Number:
233-036-2
NACRES:
NA.22

vapor pressure

14.7 hPa ( 20 °C)

Quality Level

autoignition temp.

390 °C

potency

132 mg/kg LD50, oral (Rat)

expl. lim.

4.2-32.5 % (v/v)

mp

-76 °C

transition temp

flash point >130 °C

density

1.68 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/Cl2S2/c1-3-4-2

InChI key

PXJJSXABGXMUSU-UHFFFAOYSA-N

Analysis Note

Assay (ex Cl): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.679 - 1.680

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service