Skip to Content
Merck
All Photos(1)

Documents

638A

Sigma-Aldrich

Harris′ Alum Hematoxylin Mercury Free

HARLECO® Nuclear stain used in routine H&E and Pap staining. Mercury-free formulation

Synonym(s):

H&E Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

Quality Level

form

liquid

mp

-0.1 °C

transition temp

flash point 62.2 °C

density

0.79 g/cm3

λmax

555-565 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

Application

HARLECO® Nuclear stain used in routine H&E and Pap staining. Mercury-free formulation
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Mercury-free formulation. Uses sodium iodate as an oxidizer.
Nuclear stain used in routine hematoxylin and eosin staining, as well as Pap staining.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

144.0 °F

Flash Point(C)

62.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service