Skip to Content
Merck
All Photos(1)

Key Documents

554995-M

Millipore

RGS Proteins Inhibitor II, CCG-50014

The RGS Proteins Inhibitor II, CCG-50014 controls the biological activity of RGS4 proteins. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

RGS Proteins Inhibitor II, CCG-50014, 4-(4-Fluorobenzyl)-2-p-tolyl-1,2,4-thiadiazolidine-3,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13FN2O2S
Molecular Weight:
316.35
UNSPSC Code:
12352202

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable thiadiazolidinedione compound that acts as a direct, potent, selective and cysteine-reactive irreversible inhibitor of RGS4 over other RGS proteins (IC50 = 0.03, 0.12, 3.5, 11, >200 and >200 µM against RGS4, RGS19, RGS16, RGS8, RGS7 and RGS4Cys using FCPIA assay, respectively). Shown to block RGS4-Gα0 interaction and repress Gα0-dependent membrane localization of RGS4 in HEK-293T cells with no effect on the intrinsic rate of GTP hydrolysis by Gα0. Suggested to interact with Cys107-RGS8 allosteric regulatory site and display minimal affinity towards papain (IC50 >100 µM). A reversible inhibitor of RGS Proteins, CCG-63802 is also available (Cat. No. 554993).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Blazer, L.L., et al. 2011. Biochemistry50, 3181.
Roman, D.L., et al. 2009. J. Biomol. Screening14, 610.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service