Skip to Content
Merck
All Photos(1)

Documents

5.06417

Sigma-Aldrich

SB 242084 Hydrochloride

Synonym(s):

SB 242084 Hydrochloride, 5-HT 2C receptor antagonist, SB 242084 Hydrochloride, SB-242084 Hydrochloride, SB242084 Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19ClN4O2 · xHCl
CAS Number:
Molecular Weight:
394.85 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to pale yellow
pale yellow

solubility

water: soluble

storage temp.

−20°C

InChI

1S/C21H19ClN4O2.2ClH/c1-13-10-15-7-9-26(18(15)11-17(13)22)21(27)25-16-5-6-20(24-12-16)28-19-4-3-8-23-14(19)2;;/h3-6,8,10-12H,7,9H2,1-2H3,(H,25,27);2*1H

InChI key

GCMNSEILNIPNSX-UHFFFAOYSA-N

General description

A selective, brain-penetrant 5-HT2C receptor antagonist (pKi 8.2-9.0 ), with some effects on the 5-HT2B receptor (pKi 6.8-7.0) and 5-HT2A receptor (pKi 6.1-6.8). SB 242084 has been shown to increase the effectiveness of the selective serotonin reuptake inhibitor (SSRI) class of antidepressants, and may also reduce their side effects. In animal studies, SB-242,084 produced stimulant-type activity and reinforcing effects, somewhat similar to but much weaker than cocaine or amphetamines. In vivo (ocular) IC50 values reported at 2.08 µM.

Biochem/physiol Actions

Primary Target
5-HT2C

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Manvich, D. F., et al. 2012. J Pharmacol. Exp. Ther.342, 761.
Burghardt, N. S., et al. 2007. Biol Psychiatry62, 1111.

Sharif, N. A., et al. 2007. J. Ocul. Pharmacol. Ther.23, 1.

Knight A. R., et al. 2004. Naunyn Schmiedebergs Arch. Pharmacol.370, 114.
Kennett G.A., et al. 1997. Neuropharmacol. 36, 609.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ke Liu et al.
Cellular and molecular gastroenterology and hepatology, 13(2), 483-500 (2021-09-26)
Pancreatitis is characterized by acinar cell death and persistent inflammation. Ferroptosis is a type of lipid peroxidation-dependent necrosis, which is negatively regulated by glutathione peroxidase 4. We studied how trypsin, a serine protease secreted by pancreatic acinar cells, affects the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service