506168-M
p53 Activator VII, STIMA-1
The p53 Activator VII, STIMA-1, also referenced under CAS 91634-12-7, modulates the biological activity of p53. This small molecule/inhibitor is primarily used for Activators/Inducers applications.
Synonym(s):
p53 Activator VII, STIMA-1, 2-Vinyl-1H-quinazolin-4-one, SH group Targeting and Induction of Massive Apoptosis
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 50 mg/mL
storage temp.
2-8°C
SMILES string
n1c2c(c(nc1C=C)O)cccc2
InChI
1S/C10H8N2O/c1-2-9-11-8-6-4-3-5-7(8)10(13)12-9/h2-6H,1H2,(H,11,12,13)
InChI key
VCHFWHQHIKFOIC-UHFFFAOYSA-N
General description
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service