Skip to Content
Merck
All Photos(1)

Key Documents

5.04479

Sigma-Aldrich

JNJ 16259685 - CAS 409345-29-5 - Calbiochem

Synonym(s):

JNJ 16259685 - CAS 409345-29-5 - Calbiochem, (3,4-Dihydro-2H-pyrano[2,3-b]quinolin-7-yl)-(cis-4-methoxycyclohexyl)-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23NO3
CAS Number:
Molecular Weight:
325.40
UNSPSC Code:
12352200

Assay

≥98% (TLC)

Quality Level

form

solid

potency

0.55 nM IC50
0.34 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 25 mM

storage temp.

2-8°C

General description

An extremely potent, non-competitive antagonist for mGlu receptor subtype 1 (Ki = 0.34 nM; IC50 = 0.55 nM). 400-fold more selective for mGlu1 than for mGlu5 and no interaction with mGlu2, mGlu3, mGlu4, mGlu6, AMPA or NMDA receptors (IC50 >10 µM). Centrally active following systemic administration. Widely used for studying substance abuse, learning and memory, and neural plasticity.

Biochem/physiol Actions

Primary Target
mGlu₁

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lavreysen, H., et al. 2004. Neuropharmacology.47, 961.

Steckler, T., et al. 2005. Psychopharmacology.179, 198.

Steckler, T., et al. 2005. Behav. Brain. Res.164, 52.

Mabire, D., et al. 2005. J. Med. Chem.48, 2134.
Fukunaga, I., et al. 2007. Br. J. Pharmacol.151, 870.

Knopfel, T., et al. 2007. Br. J. Pharmacol.151, 723.

Achat-Mendes, C., et al. 2012. J. Pharmacol. Exp. Ther.343, 214.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service