Skip to Content
Merck
All Photos(2)

Key Documents

328500

Sigma-Aldrich

ERO1 Inhibitor I, Erodoxin

The ERO1 Inhibitor I, Erodoxin controls the biological activity of ERO1.

Synonym(s):

ERO1 Inhibitor I, Erodoxin, 1-Bromo-5-methoxy-2,4-dinitrobenzene, Endoplasmic Reticulum Oxidase 1 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BrN2O5
CAS Number:
Molecular Weight:
277.03
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28
Product 328500 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL (Use only fresh DMSO)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

BrC(C=C(C([N+]([O-])=O)=C1)OC)=C1[N+]([O-])=O

InChI

1S/C7H5BrN2O5/c1-15-7-2-4(8)5(9(11)12)3-6(7)10(13)14/h2-3H,1H3

InChI key

SYWWRIKZBWQUGY-UHFFFAOYSA-N

General description

A cell-permeable dinitrobromobenzene compound that acts as a selective inhibitor of yeast Endoplasmic Reticulum Oxidase 1 (ERO1), but has somewhat weaker activity against mouse ERO1α (IC50 = 400 µM). Predicted to form thiol adducts and inhibits ERO1-dependent oxidation of thioredoxin-1 (Trx1) activity in vitro. Shown to cluster with genes involved in protein folding, glycosylation and cell wall biosynthesis, and delay carboxypeptidase Y (CPY) processing to the vacuolar form in wt-yeast cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze(-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Costanzo, M., et al. 2010. Science327, 425.
Blais, J.D., et al. 2010. J. Biol. Chem.285, 20993.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service