Recommended Products
manufacturer/tradename
Chemicon®
technique(s)
western blot: suitable
detection method
chemiluminescent
General description
It is widely believed that antigen-containing membranes used for Western blot experiments should never be dried. However, this is not true. The Blot Restore kit allows multiple re-use of membranes that were developed with substrate and subsequently dried. In model experiments both PVDF and nitrocellulose membranes showed excellent performance after having been dried for at least two months.
Kit contains two components. Solutioin A re-hydrates dry membranes and washes out chemiluminescent developing reagents. Solution B strips both primary and secondary (labeled) antibodies off the membrane. Treated membranes are ready for blocking and subsequent re-probing with antibodies of choice.
Use of the Blot Restore Kit offers several advantages:
1) Conservation of samples that are expensive or available only in limited quantities,
2) Analysis of a given blot using several different antibodies, e.g. subtype- or isoform-specific antibodies,
3) Re-analysis of anomalous results and confirmation with the same or a different antibody,
4) Correcting errors in incubation with the wrong antibody,
5) Cost savings in reagents and time by reusing the same blot.
Application
The Blot Restore Kit should be used only for qualitative purposes until it has been established by comparative blot analysis that stripping does not quantitatively affect a given antigen.
This product is for research use only; not for diagnostic or in vivo use.
Components
Blot Restore Solution B (10x) - (1 container, 50 mL).
Storage and Stability
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service