Skip to Content
Merck
All Photos(1)

Documents

219370

Sigma-Aldrich

Coagulation Factor VIIa, Human Plasma

Coagulation Factor VIIa, Human Plasma, is a native coagulation factor VIIa prepared from human Factor VII using human Factor XIIa, which was removed after activation.

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥25,000 PEU/mg protein (clotting assay)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native coagulation factor VIIa from human plasma prepared from human Factor VII using human Factor XIIa, which was removed after activation. Factor VIIa provides a crossover point between the extrinsic and intrinsic pathways. Prepared from human Factor VII using Human Factor XIIa.
Native coagulation factor VIIa from human plasma prepared from human Factor VII using human Factor XIIa, which was removed after activation. In the coagulation process, after complexing with tissue factor and membrane phospholipids, Factor VIIa activates Factors IX and X to the enzymatically active Factors IXa and Xa. Provides a crossover point between the extrinsic and intrinsic pathways.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 20 mM Tris-HCl, pH 7.5.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial use aliquot and freeze (-70°C).

Analysis Note

Clotting activity is determined by comparison to a standard curve produced using normal pooled human plasma

Other Notes

Broze, G.J. 1994. In Haemostasis and Thrombosis, 3rd Edition(Bloom, et al. Eds.) Churchill Livingstone, Edinburgh, p 349.
Majerus, P.W., et al. 1990. In The Pharmacological Basis of Therapeutics, 8th Edition (Gilman, A.G., et al. Eds.) Pergamon Press, New York, p 1311.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service