Skip to Content
Merck
All Photos(1)

Key Documents

14-820-M

Sigma-Aldrich

Fms (Y969C) Protein, active

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€423.00
250 μG
€3,470.00

€423.00


Estimated to ship onApril 03, 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
€423.00
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€423.00


Estimated to ship onApril 03, 2025Details


Request a Bulk Order

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

description

protein target Fms

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... CSF1R(1436)

General description

N-terminal His6-tagged, recombinant, human Fms, amino acids 538-end containing the mutation Y969C.

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-820M when ordering the 250 µg size.
Also available in 10 μg size (cat#14-820) and in bulk (cat# 14-820-K).

Physical properties

50.2kDa

Physical form

50mM Tris/HCl pH7.5, 300mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service