Select a Size
About This Item
description
protein target MRCKα
Quality Level
purified by
(Ni2+/NTA-agarose)
technique(s)
activity assay: suitable
UniProt accession no.
Gene Information
human ... CDC42BPA(8476)
General description
COFACTOR: Magnesium.
SIZE: 1732 amino acids; 197307 Da
SUBUNIT: Homodimer and homotetramer via the coiled coil regions. Interacts tightly with GTP-bound but not GDP-bound CDC42.
SUBCELLULAR LOCATION: Cytoplasm (By similarity). Note=Displays a dispersed punctate distribution and concentrates along the cell periphery, especially at the leading edge and cell-cell junction. This concentration is PH-domain dependent (By similarity).
TISSUE SPECIFICITY: Abundant in the heart, brain, skeletal muscle, kidney, and pancreas, with little or no expression in the lung and liver.
SIMILARITY: SwissProt: Q5VT25 ## Belongs to the protein kinase superfamily. AGC Ser/Thr protein kinase family. DMPK subfamily. & Contains 1 AGC-kinase C-terminal domain. & Contains 1 CNH domain. & Contains 1 CRIB domain. & Contains 1 PH domain. & Contains 1 phorbol-ester/DAG-type zinc finger. & Contains 1 protein kinase domain.
Packaging
Quality
Physical properties
Storage and Stability
Analysis Note
Disclaimer
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service