Skip to Content
Merck
All Photos(1)

Documents

1.46311

Millipore

FREY Liquid Medium

for mycoplasma, suitable for sterility testing, pkg of 100 mL (in 125 mL plastic bottle with screw cap)

Synonym(s):

Mycoplasma Broth, Mycoplasma Liquid Medium, FREY Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.75

Agency

EP 2.6.7

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

6 mo.

feature

ready-to-use

packaging

pkg of 100 mL (in 125 mL plastic bottle with screw cap)

pH

7.8 ( in H2O)

application(s)

industrial qc
microbiology
pharmaceutical

sterility testing

storage temp.

2-8°C

suitability

mycoplasma (avian)

General description

FREY Broth is supplemented with NAD to allow the growth of M. synoviae. Color change of the medium to yellow or purple indicates growth of mycoplasmas, due to Phenol Red in the medium. Penicillin inhibits the growth of accompanying bacterial flora.

Application

FREY Broth is recommended for the detection of avian mycoplasmas in pharmaceutical products, especially in bulk vaccines, cell banks, and virus cultures.

Features and Benefits

  • Ready-to-use convenience as no further preparation steps are required.
  • Pharmaceutical products such as Biopharmaceuticals, Vaccines, Cell cultures, and Virus cultures.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service