Skip to Content
Merck
All Photos(1)

Key Documents

14-553-M

Sigma-Aldrich

EphB2 Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€578.00
250 μG
€3,470.00

€578.00


Estimated to ship onApril 08, 2025Details



Select a Size

Change View
10 μG
€578.00
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€578.00


Estimated to ship onApril 08, 2025Details


biological source

human

Quality Level

mol wt

Mw 52 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

Human EphB2 expressed by baculovirus in Sf21 insect cells
N-terminal His6-tagged recombinant human EphB2 residues 560-end

Application

Active, N-terminal His6-tagged recombinant human EphB2 residues 560-end, for use in Kinase Assays.
Research Category
Inflammation & Immunology

Biochem/physiol Actions

  • Protein Target: EphB2
  • Target Sub-Family:TK

Packaging

Also available in 250 μg size --call for pricing and availability and reference catalog number 14-553M when ordering the 250 μg size.

Quality

Routinely evaluated by phosphorylation of Poly (Glu4-Tyr) (4:1)

Physical form

Format: Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service