1.24001
Spectromelt® A 22
lithium metaborate/di-lithium tetraborate 22:12
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352402
Recommended Products
Analysis Note
Chloride (Cl): ≤ 50 ppm
Phosphate (PO₄): ≤ 20 ppm
Silicate (SiO₂): ≤ 100 ppm
Sulfate (SO₄): ≤ 50 ppm
Ag (Silver): ≤ 5 ppm
Al (Aluminium): ≤ 10 ppm
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 20 ppm
Cd (Cadmium): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cr (Chromium): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 20 ppm
Ga (Gallium): ≤ 5 ppm
K (Potassium): ≤ 10 ppm
Mg (Magnesium): ≤ 5 ppm
Mn (Manganese): ≤ 5 ppm
Na (Sodium): ≤ 20 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Sr (Strontium): ≤ 5 ppm
Zn (Zinc): ≤ 10 ppm
Loss on ignition (900 °C): ≤ 0.5 %
Bulk density: about 130
Phosphate (PO₄): ≤ 20 ppm
Silicate (SiO₂): ≤ 100 ppm
Sulfate (SO₄): ≤ 50 ppm
Ag (Silver): ≤ 5 ppm
Al (Aluminium): ≤ 10 ppm
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 20 ppm
Cd (Cadmium): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cr (Chromium): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 20 ppm
Ga (Gallium): ≤ 5 ppm
K (Potassium): ≤ 10 ppm
Mg (Magnesium): ≤ 5 ppm
Mn (Manganese): ≤ 5 ppm
Na (Sodium): ≤ 20 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Sr (Strontium): ≤ 5 ppm
Zn (Zinc): ≤ 10 ppm
Loss on ignition (900 °C): ≤ 0.5 %
Bulk density: about 130
Legal Information
SPECTROMELT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service