Skip to Content
Merck
All Photos(1)

Key Documents

1.18452

Supelco

Aluminium Test Refill pack

for use with MCOLORTEST®

Synonym(s):

Aluminum Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing

Select a Size

150 TESTS
€228.00

€228.00


Estimated to ship onMarch 24, 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
150 TESTS
€228.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€228.00


Estimated to ship onMarch 24, 2025Details


Request a Bulk OrderRequest more information

feature

refill

Quality Level

specific analyte(s)

aluminium

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

This refill pack contains all the reagents for the MQuant® test kits 1.18386 and 1.14413 for the colorimetric evaluation of aluminium. When the reagents of the MQuant® test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MQuant® test kits, making it a particularly economical system.

Method
In weakly acidic, acetate-buffered solution aluminium ions react with chromazurol S to form a blue compound. Due to the intrinsic orange coloration of the reagent blank, the measurement solution is orange-red to red in color. The aluminium concentration is measured semiquantitatively by visual colorimetric evaluation.

The test can be used for ammonium determination in
  • Groundwater and surface water, seawater
  • Drinking water and mineral water
  • Waters from aquaculture
  • Boiler and boiler feed water, cooling water
  • Process water
  • Industrial water
  • Wastewater and percolating water
  • Electroplating wastewater
  • Swimming-pool water

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service