Skip to Content
Merck
All Photos(1)

Documents

1.10886

Millipore

Blood agar (base)

Synonym(s):

Blood agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

form

medium granules

Quality Level

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

6.6-7.0 (37 °C, 40 g/L in H2O, after autoclaving)

solubility

40 g/L

application(s)

clinical
food and beverages
microbiology

storage temp.

20-25°C

General description

for the isolation and cultivation of various fastidious microorganisms

Analysis Note

Appearance (clearness):clear to slightly opalescent
Appearance (colour):yellowish brown
pH-value (25 °C):6.6 - 7.0
Gel strength (Gelomat assay) (Base):≥ 50 g
Solidification behaviour (2 hrs., 40 °C):liquid
Stability test (Colour and hemolysis):passes test
Recovery on test medium (Staphylococcus aureus ATCC 25923 (WDCM 00034)):≥ 70 %
Recovery on test medium (Streptococcus pyogenes ATCC 12344):≥ 70 %
Recovery on test medium (Streptococcus agalactiae ATCC 13813):≥ 70 %
Recovery on test medium (Streptococcus pneumoniae ATCC 6301):≥ 70 %
Recovery on test medium (Listeria monocytogenes ATCC 19118):≥ 70 %
Recovery on test medium (Bacillus cereus ATCC 11778 (WDCM 00001)):≥ 70 %
Recovery on test medium (Clostridium perfringens ATCC 13124 (WDCM 00007)):≥ 70 %
Hemolysis (Streptococcus pyogenes ATCC 12344):β
Hemolysis (Streptococcus agalactiae ATCC 13813):-
Hemolysis (Streptococcus pneumoniae ATCC 6301):α
Hemolysis (Bacillus cereus ATCC 11778 (WDCM 00001)):β
Hemolysis (Clostridium perfringens ATCC 13124 (WDCM 00007)):β
Bacitracin test (Streptococcus pyogenes ATCC 12344):+
Incubation: 24 hrs.; 35 °C; aerobic (Clostridium perfringens anaerobic with Anaerocult A).

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service