Skip to Content
Merck
All Photos(2)

Key Documents

1.07570

Supelco

Extran® AP 41

greener alternative
Sign Into View Organizational & Contract Pricing

Select a Size

2 KG
€150.00
25 KG
€1,180.00

€150.00


Estimated to ship onJuly 02, 2025Details


Request a Bulk Order

Select a Size

Change View
2 KG
€150.00
25 KG
€1,180.00

About This Item

UNSPSC Code:
41103206
NACRES:
NA.21

€150.00


Estimated to ship onJuly 02, 2025Details


Request a Bulk Order

enzyme activity

0.060-0.090 IU/g

Quality Level

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

11.4 (20 °C, 3 g/L in H2O)

bulk density

750 kg/m3

greener alternative category

storage temp.

no temp limit

General description

This product is identical to EX0992 with the same quality and specification. Some pack sizes have changed to provide a globally harmonized offering.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is environmentally friendly having bio-degradable active ingredients. Click here for more information.

Analysis Note

pH-value (1 %; water): 11 - 12
Foaming volume (5 %; water): ≤ 1 ml
Bulk density: 65 - 80 g/100 ml
Enzymeactivity: 0.060 - 0.090 Anson U/g

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service