Skip to Content
Merck
All Photos(1)

Key Documents

1.02914

Sigma-Aldrich

Nitromethane-D3

deuteration degree min. 99% for NMR spectroscopy MagniSolv

Synonym(s):

Nitromethane-D3, Trideuteronitromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CD3NO2
CAS Number:
Molecular Weight:
64.06
MDL number:
UNSPSC Code:
12142201
EC Index Number:
235-892-2

vapor pressure

36 hPa ( 20 °C)

form

liquid

autoignition temp.

415 °C

potency

940 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7.1-63 % (v/v)

technique(s)

NMR: suitable

impurities

≤0.03% Water (H<SUB>2</SUB>O) (NMR)

mp

-26 °C

transition temp

flash point 44 °C

solubility

117 g/L

density

1.19 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/CH3NO2/c1-2(3)4/h1H3/i1D3

InChI key

LYGJENNIWJXYER-FIBGUPNXSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Analysis Note

Deuteration degree: ≥ 99 %
Water (NMR, H₂O): ≤ 0.03 %
Performance test (NMR-spectrum): passes test

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service