Skip to Content
Merck
All Photos(2)

Key Documents

09-872

Sigma-Aldrich

Anti-Methylcytosine dioxygenase TET1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

tet oncogene 1, Ten-eleven translocation 1 gene protein, CXXC-type zinc finger protein 6, methylcytosine dioxygenase TET1, CXXC finger 6, ten-eleven translocation-1, Leukemia-associated protein with a CXXC domain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

ChIP: suitable (ChIP-seq)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... TET1(80312)

General description

Methylcytosine dioxygenase TET1 (mTET1) is a member of the TET family and contains a characteristic CXXC-type zinc finger domain. mTET1 binds methylcytosine (5mC), a minor, mammalian DNA base necessary to development and retrotransposon silencing, and acts as a catalyst to induce the conversion of 5mC to 5-hydromethylcytosine (hmC or 5hmC), a mammalian DNA base thought to be involved in chromatin structure and in the development of fetal brain, heart and lung tissues. Recent studies indicate that mTET1 regulates the levels of 5hmC, thereby influencing patterns in DNA methylation.

Specificity

Other homologies: Rat (66% sequence homology), Human (61% sequence homology).
This antibody recognizes methylcytosine dioxygenase TET1 near the C-terminus.

Immunogen

Epitope: Near the C-terminus
KLH-conjugated linear peptide corresponding to mouse Methylcytosine dioxygenase TET1 near the C-terminus.

Application

Anti-Methylcytosine dioxygenase TET1 Antibody is a Rabbit Polyclonal Antibody for detection of Methylcytosine dioxygenase TET1 also known as tet oncogene 1 & has been validated in WB, ChIP-seq.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Western Blot (Snap i.d.) Analysis: 5 µg/mL from a previous lot detected Methylcytosine dioxygenase TET1 in 10 µg of mouse embryonic stem cell lysate.

Quality

Evaluated by Western Blot in mouse embryonic stem cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Methylcytosine dioxygenase TET1 in 10 µg of mouse embryonic stem cell lysate.

Target description

~220 kDa observed.
Uncharacterized bands may be seen at ~32, 68, and 70 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse embryonic stem cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Vella et al.
Molecular cell, 49(4), 645-656 (2013-01-29)
O-linked N-acetylglucosamine (O-GlcNAc) transferase (Ogt) activity is essential for embryonic stem cell (ESC) viability and mouse development. Ogt is present both in the cytoplasm and the nucleus of different cell types and catalyzes serine and threonine glycosylation. We have characterized
Suppression of TET1-dependent DNA demethylation is essential for KRAS-mediated transformation.
Wu, BK; Brenner, C
Cell Reports null
Dynamic hydroxymethylation of deoxyribonucleic acid marks differentiation-associated enhancers.
Serandour, Aurelien A, et al.
Nucleic Acids Research (2012)
Naoki Okashita et al.
Development (Cambridge, England), 141(2), 269-280 (2013-12-18)
Ten-eleven translocation (TET) proteins oxidize 5-methylcytosine (5mC) to 5-hydroxymethylcytosine (5hmC), 5-formylcytosine (5fC) and 5-carboxylcytosine (5caC). 5fC and 5caC can be excised and repaired by the base excision repair (BER) pathway, implicating 5mC oxidation in active DNA demethylation. Genome-wide DNA methylation
Alain R Weber et al.
Nature communications, 7, 10806-10806 (2016-03-05)
Cytosine methylation in CpG dinucleotides is an epigenetic DNA modification dynamically established and maintained by DNA methyltransferases and demethylases. Molecular mechanisms of active DNA demethylation began to surface only recently with the discovery of the 5-methylcytosine (5mC)-directed hydroxylase and base

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service