Skip to Content
Merck
All Photos(1)

Key Documents

KEY465202486

Key Organics / BIONET

4-(4-fluorobenzoyl)piperidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
243.71
MDL number:

mp

222-226

Iupac Name

(4-fluorophenyl)(4-piperidinyl)methanone hydrochloride

Inchi Code

1S/C12H14FNO.ClH/c13-11-3-1-9(2-4-11)12(15)10-5-7-14-8-6-10;/h1-4,10,14H,5-8H2;1H

InChI key

GPKDBZQZPNOBGM-UHFFFAOYSA-N

storage temp.

Store under inert gas, Hygroscopic

Purity

90%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/LF-0526

Melting Point

222-226

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/LF-0526


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library