KEY465195804
Key Organics / BIONET
3-[(2E)-2-[(4-methylphenyl)methylidene]hydrazin-1-yl]-1,2-dihydroquinoxalin-2-one
About This Item
Iupac Name
4-methylbenzaldehyde (3-oxo-3,4-dihydro-2-quinoxalinyl)hydrazone
Inchi Code
1S/C16H14N4O/c1-11-6-8-12(9-7-11)10-17-20-15-16(21)19-14-5-3-2-4-13(14)18-15/h2-10H,1H3,(H,18,20)(H,19,21)/b17-10+
InChI key
CARPROJTONAMLC-LICLKQGHSA-N
Purity
Country of Origin
Physical Form
Product Link
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/4F-965
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.