ENAH11DD0DAB
Enamine
methyl 2-hydroxy-2-(pyrimidin-5-yl)acetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl hydroxy(5-pyrimidinyl)acetate
Inchi Code
1S/C7H8N2O3/c1-12-7(11)6(10)5-2-8-4-9-3-5/h2-4,6,10H,1H3
InChI key
GQONMURQTNGONL-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
oil
Product Link
https://www.enaminestore.com/catalog/EN300-1602001
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-1602001
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.