ENA341296982
Enamine
ethyl 2-(4-bromobenzenesulfonyl)acetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
49-50
Iupac Name
ethyl [(4-bromophenyl)sulfonyl]acetate
Inchi Code
1S/C10H11BrO4S/c1-2-15-10(12)7-16(13,14)9-5-3-8(11)4-6-9/h3-6H,2,7H2,1H3
InChI key
BHWNAYXWMDZNMN-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-94433
Melting Point
49-50
MSDS
https://www.enaminestore.com/catalog/EN300-94433
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.