Skip to Content
Merck
All Photos(1)

Key Documents

COM945365449

Combi-Blocks, Inc.

N-(5-Bromo-2-cyanophenyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
239.07
MDL number:

Iupac Name

N-(5-bromo-2-cyanophenyl)acetamide

Inchi Code

1S/C9H7BrN2O/c1-6(13)12-9-4-8(10)3-2-7(9)5-11/h2-4H,1H3,(H,12,13)

InChI key

QJRJQXQWVUGHFM-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QL-4331

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QL-4331


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library