COM633414142
Combi-Blocks, Inc.
2-Methoxy-4-(tributylstannyl)thiazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 4-(tributylstannyl)-1,3-thiazol-2-yl ether
Inchi Code
1S/C4H4NOS.3C4H9.Sn/c1-6-4-5-2-3-7-4;3*1-3-4-2;/h3H,1H3;3*1,3-4H2,2H3;
InChI key
JCZRGWAGRPABLT-UHFFFAOYSA-N
storage temp.
Refrigerated
Purity
96%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?SS-0787
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?SS-0787
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.