Skip to Content
Merck
All Photos(1)

Key Documents

870709P

Avanti

04:1 Coenzyme A

Avanti Research - A Croda Brand 870709P, powder

Synonym(s):

(E)-but-2-enoyl Coenzyme A (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H37N7Na3O17P3S
CAS Number:
Molecular Weight:
901.55
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870709P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870709P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(/C=C/C)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[Na+].[Na+].[Na+]

General description

04:1 Coenzyme A, also known as crotonyl‐CoA, is produced from reversible dehydration of 4‐hydroxybutyryl coenzyme A by the enzyme 4‐hydroxybutyryl‐CoA dehydratase. The conversion of crotonyl-CoA to butyryl-CoA during the butyrate fermentation in the human gut releases the extra energy such as potential of mean force (PMF).

Packaging

5 mL Amber Glass Screw Cap Vial (870709P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The complete stereochemistry of the enzymatic dehydration of 4-hydroxybutyryl coenzyme A to crotonyl coenzyme A.
Peter Friedrich et al.
Angewandte Chemie (International ed. in English), 47(17), 3254-3257 (2008-03-19)
Bacterial energy metabolism
Molecular Medicine, 47(17), 201-233 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service