Skip to Content
Merck
All Photos(1)

Key Documents

ATE428009119

AstaTech, Inc.

9-BROMO-2-HYDROXY-7-METHYL-4H-PYRIDO[1,2-A]PYRIMIDIN-4-ONE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
255.07
MDL number:

Iupac Name

9-bromo-2-hydroxy-7-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

Inchi Code

1S/C9H7BrN2O2/c1-5-2-6(10)9-11-7(13)3-8(14)12(9)4-5/h2,4H,3H2,1H3

InChI key

MSMTVXPWEYVJPQ-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

98%

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=43472

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=32295


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library