ATE398812728
AstaTech, Inc.
METHYL 4-CHLORO-6-(TRIFLUOROMETHYL)PICOLINATE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 4-chloro-6-(trifluoromethyl)-2-pyridinecarboxylate
Inchi Code
1S/C8H5ClF3NO2/c1-15-7(14)5-2-4(9)3-6(13-5)8(10,11)12/h2-3H,1H3
InChI key
QGNOZTMZBPOLGT-UHFFFAOYSA-N
storage temp.
2-8 C
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
White to Yellow Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=19144
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=82655
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.