Skip to Content
Merck
All Photos(1)

Key Documents

APO455836522

Apollo Scientific Ltd

3,4,5-Trifluorostyrene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
158.12
MDL number:
Pricing and availability is not currently available.

Iupac Name

1,2,3-trifluoro-5-vinylbenzene

Inchi Code

1S/C8H5F3/c1-2-5-3-6(9)8(11)7(10)4-5/h2-4H,1H2

InChI key

SBZYGHMGBOZTOU-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid-Clear

Product Link

https://store.apolloscientific.co.uk/product/345-trifluorostyrene

Synonym

5-Ethenyl-1,2,3-trifluorobenzene, 1,2,3-Trifluoro-5-vinylbenzene

Pictograms

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/345-trifluorostyrene


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library