AOB640091986
AOBChem
4-Hydroxy-2-(trifluoromethyl)phenylboronic acid
About This Item
Iupac Name
4-hydroxy-2-(trifluoromethyl)phenylboronic acid
Inchi Code
1S/C7H6BF3O3/c9-7(10,11)5-3-4(12)1-2-6(5)8(13)14/h1-3,12-14H
InChI key
MHKOVSRFDDLIFU-UHFFFAOYSA-N
Purity
Country of Origin
Physical Form
Product Link
MSDS
https://aobchem.com/pub/media/files/sds/12099.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.