Skip to Content
Merck
All Photos(1)

Documents

AMBH93E4C1E8

Ambeed, Inc.

2-Hydrazinyl-5-nitropyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
154.13
MDL number:

Iupac Name

2-hydrazino-5-[hydroxy(oxido)amino]pyridine

Inchi Code

1S/C5H6N4O2/c6-8-5-2-1-4(3-7-5)9(10)11/h1-3H,6H2,(H,7,8)

InChI key

DDWAPSXNXHYQLK-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,Store in freezer, under -20C

Purity

95%

Country of Origin

CN

Shipping Temperature

Shipped in cold pack

Physical Form

solid

Product Link

https://www.ambeed.com/products/6343-98-2.html

Synonym

2-Hydrazono-5-nitro-1,2-dihydropyridine

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

MSDS

https://www.ambeed.com/products/6343-98-2.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service