Z760307
Ohaus® MB-23 and MB-25 moisture analyzers
Model MB23, AC/DC input 230 V AC, European 2-pin plug
Synonym(s):
moisture meters
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Ohaus® 80252470 EU
weighing capacity measuring range
110 g
parameter
50-160 °C temp. limit
W × H × depth
17 cm × 13 cm × 28 cm
platform diam.
90 mm
General description
The MB23 has an IR heating element. The non-glass infrared heating element provides even heating and is ideal for food testing, textiles, wastewater, and ceramics applications.
The Moisture analyzers combine high quality construction and easy setup. The LCD display is backlit, and it has a standard RS232 port.
- easy to clean design
- heating chamber is low maintenance
- compact footprint
Legal Information
Ohaus is a registered trademark of Ohaus Corp.