Documents
Z744036
Work station anti-static ionizer for Plas-Labs glove box
AC/DC input 100 V, 50/60 Hz, Japan plug
Synonym(s):
Work station anti-static ionizer for Plas-Labs glove box
About This Item
UNSPSC Code:
42132201
NACRES:
NB.79
material
electrode (Thoriated tungsten)
manufacturer/tradename
Plas-Labs 800AS/SPI/J
Overall H × W × depth
5.0 in. × 4 1/4 in. × 2 1/2 in.
weight
2 lb
General description
The anti-static ionizer effectively eliminates all static charges within 36 inches of the unit and is useful in any Plas-Labs glove box or isolation chamber.
Operation is simple; place the unit on a flat surface and turn the power on.
Unit features include: ion pulse rate adjustment from 1 to 5 pulses/second, ion balance adjustment and a compact footprint. The unit is virtually maintenance free; the emitter points should be cleaned when a visual accumulation of dust or powder occurs.