Key Documents
Z301302
Quickfit® plain bends
joint: ST/NS 14/23
Synonym(s):
RA9/11
Select a Size
1 EA
€44.70
€44.70
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€44.70
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€44.70
Please contact Customer Service for Availability
Joint
joint: ST/NS14/23
manufacturer/tradename
Quickfit RA 9/11
availability
not available in USA and Canada
not available in USA and Canada
General description
Socket to cone. Generally used as distilling adapters.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)