Key Documents
Z179558
Supelco
Pre-column for Michel-Miller columns
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
manufacturer/tradename
Ace Glass 579634
technique(s)
LPLC: suitable
L × I.D.
130 mm × 22 mm
General description
For the addition of pre-purified sample mixtures to main column. Technique allows silica gel or alumina-packed columns to be reused several times. Suitable for use in final stages of slurry packing; stacks on top of main M-M column using coupling. Connections are Ace Thred #11.
Choose from one of the most recent versions:
Certificates of Analysis (COA)