W509434
2-Butanethiol
≥98%
Synonym(s):
sec-Butyl mercaptan, Mercaptan C4
About This Item
Recommended Products
biological source
synthetic
vapor pressure
142 mmHg ( 37.7 °C)
Assay
≥98%
refractive index
n20/D 1.436 (lit.)
bp
84.6-85.2 °C (lit.)
density
0.83 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Organoleptic
sulfurous
SMILES string
CCC(C)S
InChI
1S/C4H10S/c1-3-4(2)5/h4-5H,3H2,1-2H3
InChI key
LOCHFZBWPCLPAN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-9.4 °F - closed cup
Flash Point(C)
-23 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service